missing translation for 'onlineSavingsMsg'
Learn More

Nogo-A Antibody (387521) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30731976 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731976 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731976 Supplier Novus Biologicals Supplier No. FAB3098JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Nogo-A Monoclonal antibody specifically detects Nogo-A in Rat samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nogo-A
Applications Immunohistochemistry
Classification Monoclonal
Clone 387521
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias ASY;Nbla00271;Nbla10545;NI220/250;Nogo;NSP;NSP-CL;r;Reticulon-4;Rtn4;RTN4;RTN4-A;RTN4-B1;RTN4-B2;RTN4-C;RTN-X
Host Species Mouse
Immunogen KLH-coupled rat Nogo-A synthetic peptide, SYDSIKLEPENPPPYEEA (aa 623-640), Accession # Q9JK11
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 57142
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.