Learn More
Noggin Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-40413PCP
This item is not returnable.
View return policy
Description
Noggin Polyclonal antibody specifically detects Noggin in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Noggin | |
| PerCP | |
| Store at 4C in the dark. |
| Western Blot | |
| Western Blot |