missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Noggin Antibody (2C10), Novus Biologicals™

Product Code. 18347188 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347188 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347188 Supplier Novus Biologicals™ Supplier No. H00009241M11

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Noggin Monoclonal antibody specifically detects Noggin in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Noggin
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2C10
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100-1:2000
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005441
Gene Alias noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1
Host Species Mouse
Immunogen NOG (NP_005441, 27 a.a. ∽ 145 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GQHYLHIRPAPSDNLPLVDLIEHPDPIFDPKEKDLNETLLRSLLGGHYDPGFMATSPPEDRPGGGGGAAGGAEDLAELDQLLRQRPSGAMPSEIKGLEFSEGLAQGKKQRLSKKLRRKL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9241
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.