missing translation for 'onlineSavingsMsg'
Learn More

Nodal Rabbit anti-Human, Rat, Clone: S06-5A3, Novus Biologicals™

Product Code. 18335866
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335866 25 μg 25µL
18359251 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18335866 Supplier Novus Biologicals Supplier No. NBP31506825UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Nodal Monoclonal antibody specifically detects Nodal in Human, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nodal
Applications Western Blot
Classification Monoclonal
Clone S06-5A3
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias MGC138230, nodal homolog, nodal homolog (mouse), nodal, mouse, homolog
Host Species Rabbit
Immunogen A synthetic peptide of human Nodal (Uniprot#: Q96S42)
Purification Method Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cytokine Research, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4838
Target Species Human, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.