missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NOD4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12849162
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12849162 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12849162 Supplier Invitrogen™ Supplier No. PA521017

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is EL4 cell lysate. PA5-21017 can be used with blocking peptide PEP-1131.

NOD4 is a member of the NOD (nucleotide-binding oligomerization domain) family, a group of proteins that are involved in innate immune defense. NOD4 contains a CARD-like domain, a central NOD domain and a large LRR region. NOD4, an IFN-gamma-inducible nuclear protein, plays a role in homeostatic control of innate immunity and in antiviral defense mechanisms. As a key negative regulator of NF-kappa-B and type I interferon signaling, NOD4 may be a useful target for manipulating immune responses against infectious or inflammation-associated diseases, including cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOD4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene NLRC5
Gene Accession No. C3VPR6, Q86WI3
Gene Alias AI451557; AK220210; Caterpiller protein 16.1; CLR16.1; copine II; Cpne2; NLR family CARD domain containing 5; NLR family, CARD domain containing 5; Nlrc5; NOD27; NOD4; NOD-like receptor C5; Nucleotide-binding oligomerization domain protein 27; Nucleotide-binding oligomerization domain protein 4; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 5; nucleotide-binding oligomerization domains 27; protein NLRC5
Gene Symbols NLRC5
Host Species Rabbit
Immunogen An 18 amino acid peptide near the amino terminus of human NOD4.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 434341, 84166
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.