missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NOD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565491
This item is not returnable.
View return policy
Description
Immunogen sequence: SNDSRIQRHRK ALLSKVGGGP ELGGPWHRLA SLLLVEGLTD LQLREHDFTQ VEATRGGGHP ARTVALDRLF LPLSRVSVPP RVSITIGV Highest antigen sequence identity to the following orthologs - mouse 79%, rat 74%.
NOD3 is a member of the NOD (nucleotide-binding oligomerization domain) family, a group of proteins that are involved in innate immune defense. NOD3 also known as NLR family CARD containing 3 (NLRC3), is predominantly expressed in the immune system, particularly in T lymphocytes, and its expression is strongly down-regulated following stimulation of the T-cell receptor complex and CD28, suggesting that NOD3 plays a role in attenuating the activation of T cells. NOD3 inhibits NF-βB, AP-1 and NFAT transcriptional activation in Jurkat T cells downstream of CD3/CD28 stimulation or treatment with PMA/ionomycin and decreases IL-2 and CD25 mRNA induction in activated cells.
Specifications
| NOD3 | |
| Polyclonal | |
| Unconjugated | |
| NLRC3 | |
| CARD15-like protein; caterpillar 16.2; Caterpiller 16.2; caterpiller protein 16.2; CLR16.2; D230007K08Rik; mFLJ00348; NACHT, LRR and CARD domains-containing protein 3; NLR family CARD domain containing 3; NLR family CARD domain-containing protein 3; NLR family, CARD domain containing 3; NLR family, CARD domain containing 3, pseudogene 1; Nlrc3; Nlrc3-ps1; NOD3; NOD-like receptor C3; Nucleotide-binding oligomerization domain protein 3; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 3; protein NLRC3; RGD1304902 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 197358 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7RTR2 | |
| NLRC3 | |
| Recombinant Human NOD3. Recombinant protein control fragment (Product #RP-104842). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction