missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Gene ID (Entrez) | 64127 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP2-27118. For further blocking peptide related protocol, click here . |
| Gene Alias | BLAU, CARD15caspase recruitment domain protein 15, caspase recruitment domain family, member 15, Caspase recruitment domain-containing protein 15, CDACUG, CLR16.3, IBD1NLR family, CARD domain containing 2, Inflammatory bowel disease protein 1, NLRC2, NOD-like receptor C2, nucleotide-binding oligomerization domain 2, nucleotide-binding oligomerization domain containing 2, nucleotide-binding oligomerization domain, leucine rich repeat and CARD domaincontaining 2, nucleotide-binding oligomerization domain-containing protein 2, PSORAS1NOD2B |
| Gene Symbol | NOD2 |
| Quantity | 0.1 mg |
| Research Category | Apoptosis, Autophagy, Cancer, Caspases, Cell Biology |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?