missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOD2 Antibody (2D9), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-524PE
This item is not returnable.
View return policy
Description
NOD2 Monoclonal specifically detects NOD2 in Human, Mouse samples. It is validated for Flow Cytometry, Flow (Intracellular).
Specifications
| NOD2 | |
| Monoclonal | |
| PE | |
| Q9HC29 | |
| NOD2 | |
| Recombinant human NOD2 protein corresponding to residues 28-301. [UniProt# Q9HC29] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human and mouse. Immunogen has 92% homology to primate. | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 2D9 | |
| Flow Cytometry, Flow (Intracellular) | |
| BLAU, CARD15caspase recruitment domain protein 15, caspase recruitment domain family, member 15, Caspase recruitment domain-containing protein 15, CDACUG, CLR16.3, IBD1NLR family, CARD domain containing 2, Inflammatory bowel disease protein 1, NLRC2, NOD-like receptor C2, nucleotide-binding oligomerization domain 2, nucleotide-binding oligomerization domain containing 2, nucleotide-binding oligomerization domain, leucine rich repeat and CARD domaincontaining 2, nucleotide-binding oligomerization domain-containing protein 2, PSORAS1NOD2B | |
| Mouse | |
| 110 kDa | |
| 0.1 mL | |
| Apoptosis, Autophagy, Cancer, Caspases, Cell Biology | |
| 64127 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction