missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOD2 Antibody (2D9) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80883
This item is not returnable.
View return policy
Description
NOD2 Monoclonal specifically detects NOD2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF-ready.
Specifications
| NOD2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 1:100-1:2000, Flow Cytometry reported in scientific literature (PMID 28241127), Immunohistochemistry : 1:10-1:500, Immunocytochemistry/Immunofluorescence : 1:50-1:500, Immunoprecipitation : 1:10-1:500, Immunohistochemistry-Paraffin : 1:10-1:500, Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF-ready | |
| BLAU, CARD15caspase recruitment domain protein 15, caspase recruitment domain family, member 15, Caspase recruitment domain-containing protein 15, CDACUG, CLR16.3, IBD1NLR family, CARD domain containing 2, Inflammatory bowel disease protein 1, NLRC2, NOD-like receptor C2, nucleotide-binding oligomerization domain 2, nucleotide-binding oligomerization domain containing 2, nucleotide-binding oligomerization domain, leucine rich repeat and CARD domaincontaining 2, nucleotide-binding oligomerization domain-containing protein 2, PSORAS1NOD2B | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF | |
| 2D9 | |
| Unconjugated | |
| PBS | |
| NOD2 | |
| Recombinant human NOD2 protein corresponding to residues 28-301. [UniProt# Q9HC29] | |
| 0.1 mL | |
| Apoptosis, Autophagy, Cancer, Caspases, Cell Biology | |
| 64127 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction