missing translation for 'onlineSavingsMsg'
Learn More

NOD1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18423986 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423986 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423986 Supplier Novus Biologicals Supplier No. NBP176286JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NOD1 Polyclonal antibody specifically detects NOD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOD1
Applications Western Blot, Flow Cytometry, ELISA
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias CARD4NLR family, CARD domain containing 1, caspase recruitment domain family, member 4, Caspase recruitment domain-containing protein 4, CLR7.1, NLRC1, nucleotide-binding oligomerization domain containing 1, nucleotide-binding oligomerization domain, leucine rich repeat and CARD domaincontaining 1, nucleotide-binding oligomerization domain-containing protein 1
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the carboxy end of human NOD1. The immunogen is located within the last 50 amino acids of NOD1. Amino Acid Squence: NLIKEPEEAKVYEDEKR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Asthma, Autophagy, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10392
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.