Learn More
Nocodazole, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
- Used in cell division & replication research, cancer research, microscopy, and flow cytometry
- Nocodazole is usually used at a concentration of 40–100 ng/mL of culture medium for a duration of 12 to 18 hours
- Prolonged arrest of cells in mitosis due to nocodazole treatment typically results in cell death by apoptosis
Specifications
Specifications
| Quantity | 10 mg |