missing translation for 'onlineSavingsMsg'
Learn More

Nocodazole, MedChemExpress

Product Code. 16425728
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16425728

Brand: Medchem Express HY13520

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nocodazole (Oncodazole) is a rapidly-reversible inhibitor of microtubule. Nocodazole binds to β-tubulin and disrupts microtubule assembly/disassembly dynamics, which prevents mitosis and induces apoptosis in tumor cells. Nocodazole inhibits Bcr-Abl, and activates CRISPR/Cas9.

Nocodazole (Oncodazole) is a rapidly-reversible inhibitor of microtubule. Nocodazole binds to β-tubulin and disrupts microtubule assembly/disassembly dynamics, which prevents mitosis and induces apoptosis in tumor cells. Nocodazole inhibits Bcr-Abl, and activates CRISPR/Cas9.

Specifications

Chemical Name or Material Nocodazole
Physical Form Solid
CAS 31430-18-9
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C14H11N3O3S
For Use With (Application) Cancer-Kinase/protease
Synonym Oncodazole R17934
Solubility Information DMSO : 20 mg/mL (66.37 mM; Need ultrasonic)
SMILES O=C(OC)NC1=NC2=CC=C(C(C3=CC=CS3)=O)C=C2N1
Molecular Weight (g/mol) 301.32
Formula Weight 301.32
Percent Purity 98.63%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.