missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nociceptin Polyclonal Antibody
GREENER_CHOICE

Product Code. 10822851
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10822851 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10822851 Supplier Invitrogen™ Supplier No. PA3204

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA3-204 detects Nociceptin in human, rat and mouse samples. PA3-204 has successfully been used in immuncytochemical and immunohistochemical (paraffin) procedures. The PA3-204 immunogen is a synthetic peptide corresponding to the amino acids FGGFTGARKSARKLANQ of Nociceptin.

Nociceptin is the ligand of the opioid receptor-like receptor (OPRL1). It may act as a transmitter in the brain by modulating nociceptive and locomotor behavior. May be involved in neuronal differentiation and development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nociceptin
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.02% sodium azide
Gene PNOC
Gene Accession No. Q13519, Q62923, Q64387
Gene Alias N/OFQ; N23K; N23K/N27K; neuropeptide nociceptin 1; nociceptin; Nocistatin; NOP; Npnc1; OFQ; OFQ Orphanin FG; OFQ/N; ORL1 receptor agonist; Orphanin FQ; Orphanin FQ2; Pnoc; ppN/OFQ; PPNOC; pre-pro-N/OFQ; prepronociceptin; Prepronociceptin (neuropeptide nociceptin) (N23K); pronociceptin; proorphanin
Gene Symbols PNOC
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the amino acids FGGFTGARKSARKLANQ of Nociceptin.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18155, 25516, 5368
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.