missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOC4L Mouse anti-Human, Clone: PCRP-NOC4L-1E3, Novus Biologicals™
Mouse Monoclonal Antibody
£196.00
Specifications
| Antigen | NOC4L |
|---|---|
| Clone | PCRP-NOC4L-1E3 |
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array |
| Applications | Flow Cytometry, Immunofluorescence, Peptide Array |
Description
NOC4L Monoclonal specifically detects NOC4L in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
| NOC4L | |
| 0.2 mg/ml | |
| Flow Cytometry, Immunofluorescence, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| MGC3162, NET49, NOC4, NOC4 protein homolog, NOC4-like protein, nucleolar complex associated 4 homolog (S. cerevisiae), nucleolar complex protein 4 homolog, Nucleolar complex-associated protein 4-like protein | |
| Recombinant full-length human NOC4L protein | |
| Primary | |
| Store at 4°C. |
| PCRP-NOC4L-1E3 | |
| Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 79050 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title