missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOC4L Antibody (PCRP-NOC4L-1B2), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14031MFV500
This item is not returnable.
View return policy
Description
NOC4L Monoclonal antibody specifically detects NOC4L in Human samples. It is validated for Flow Cytometry
Specifications
| NOC4L | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 79050 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| PCRP-NOC4L-1B2 | |
| Flow Cytometry | |
| MGC3162, NET49, NOC4, NOC4 protein homolog, NOC4-like protein, nucleolar complex associated 4 homolog (S. cerevisiae), nucleolar complex protein 4 homolog, Nucleolar complex-associated protein 4-like protein | |
| Recombinant full-length human NOC4L protein (Uniprot: Q9BVI4) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction