missing translation for 'onlineSavingsMsg'
Learn More

NOC4L Antibody (PCRP-NOC4L-1B2), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18347632
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347632 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347632 Supplier Bio-Techne Supplier No. NBP314031AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NOC4L Monoclonal antibody specifically detects NOC4L in Human samples. It is validated for Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOC4L
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone PCRP-NOC4L-1B2
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias MGC3162, NET49, NOC4, NOC4 protein homolog, NOC4-like protein, nucleolar complex associated 4 homolog (S. cerevisiae), nucleolar complex protein 4 homolog, Nucleolar complex-associated protein 4-like protein
Host Species Mouse
Immunogen Recombinant full-length human NOC4L protein (Uniprot: Q9BVI4)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79050
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.