missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NOC3L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566939
This item is not returnable.
View return policy
Description
Immunogen sequence: DDLQLMKDLGQ RVSFLTRDLS SSEPVHAKKR KHERIIDKYE KIPRTLQTAP EKELIHLLPI KDKSGIIPQT REKPV Highest antigen sequence identity to the following orthologs - mouse 89%, rat 89%.
GADD 153, a growth arrest and DNA damage-inducible gene, encodes a C/EBP-related nuclear protein. This protein has also been designated C/EBP-homologous protein (CHOP-10 or C/EBP zeta). GADD 153 expression is induced by a variety of cellular stresses, inducing nutrient deprivation and metabolic perturbations. GADD 153 functions to block cells in G1 to S phase during cell cycle progression and acts by dimerizing with other C/EBP proteins to direct GADD 153 dimers away from classical C/EBP binding sites, recognizing instead unique nonclassical sites. Thus, GADD 153 acts as a negative modulator of C/EBP-like proteins in certain terminally differentiated cells. GADD 153 belongs to the CBF/MAK21 family, which also includes NOC2L, NOC3L and NOC4L. NOC3L, also designated factor for adipocyte differentiation 24 or Fad24, promotes adipogenesis by controlling DNA replication during the early stages of mitotic clonal expansion (MCE).
Specifications
| NOC3L | |
| Polyclonal | |
| Unconjugated | |
| NOC3L | |
| AD24; AF233884; C10orf117; factor for adipocyte differentiation 24; Fad24; NOC3 like DNA replication regulator; NOC3 protein homolog; NOC3L; NOC3L DNA replication regulator; NOC3-like DNA replication regulator; NOC3-like protein; nucleolar complex associated 3 homolog; nucleolar complex associated 3 homolog (S. cerevisiae); nucleolar complex protein 3 homolog; Nucleolar complex-associated protein 3-like protein; RGD1560656 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64318 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WTT2 | |
| NOC3L | |
| Recombinant Human NOC3L. Recombinant protein control fragment (Product #RP-105433). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction