missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NOC2L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101730
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NOC2L.
INHAT (inhibitor of acetyltransferases) complex binds to histones and masks them from being substrate for acetyltransferase. Endogenous INHAT subunits, which include the Set/TAF-Ibeta oncoprotein, associate with chromatin in vivo and can block coactivator mediated transcription when transfected in cells.
Specifications
| NOC2L | |
| Polyclonal | |
| Unconjugated | |
| NOC2L | |
| AA410003; AF155546; DKFZp564C186; FLJ35172; hypothetical protein LOC508638; NET15; NET7; NIR; NOC2 like nucleolar associated transcriptional repressor; NOC2L; NOC2L nucleolar associated transcriptional repressor; NOC2-like; NOC2-like nucleolar associated transcriptional repressor; NOC2-like protein; novel INHAT (inhibitor of histone acetyltransferase) repressor; novel INHAT repressor; nucleolar complex associated 2 homolog; nucleolar complex associated 2 homolog (S. cerevisiae); nucleolar complex protein 2 homolog; PPP1R112; Protein NOC2 homolog; protein phosphatase 1, regulatory subunit 12; RGD1309387 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26155, 313777, 57741 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9WV70, Q9Y3T9 | |
| NOC2L | |
| A synthesized peptide derived from human NOC2L(Accession Q9Y3T9), corresponding to amino acid residues V601-R651. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction