missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOBOX Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543056
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: EELLRGSAPH AQDTQSEELP PSCTISGEKK PPAVSGEATG ADAGRLCPPP Sequence homology: Human: 100%; Rabbit: 77%
NOBOX Polyclonal antibody specifically detects NOBOX in Human samples. It is validated for Western Blot.Specifications
NOBOX | |
Polyclonal | |
Unconjugated | |
NOBOX | |
Homeobox protein NOBOX; homeodomain-containing protein OG-2; LOW QUALITY PROTEIN: homeobox protein NOBOX; newborn ovary homeobox protein; newborn ovary homeobox-encoding; Nobox; NOBOX oogenesis homeobox; OG2; OG-2; OG2 homeobox; Og2x; Oocyte-specific homeobox protein; POF5; TCAG_12042 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
135935 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O60393 | |
NOBOX | |
Synthetic peptide directed towards the N-terminal of human NOBOX | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |