missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Noble Metals Standard: Au, Ir, Os, Pd, Pt, Re, Rh, Ru @ 100 μg/mL in 20% HCl, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
Noble Metals Standard: Au, Ir, Os, Pd, Pt, Re, Rh, Ru @ 100 μg/mL in 20% HCl
Specifications
Specifications
| Chemical Name or Material | Gold/Iridium/Osmium/Palladium/Platinum/Rhenium/Rhodium/Ruthenium |
| CAS | 7440-57-5, 7439-88-5, 4-2-7440, 5-3-7440, 6-4-7440, 7440-15-5 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Gold/Iridium/Osmium/Palladium/Platinum/Rhenium/Rhodium/Ruthenium |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HCl |
| Quantity | 500 mL |
| Concentration or Composition (by Analyte or Components) | Gold 100 μg/mL/Iridium 100 μg/mL/Osmium 100 μg/mL/Palladium 100 μg/mL/Platinum 100 μg/mL/Rhenium 100 μg/mL/Rhodium 100 μg/mL/Ruthenium 100 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction