missing translation for 'onlineSavingsMsg'
Learn More

Nobiletin, TRC

Product Code. 30470854
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30470854 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30470854 Supplier Toronto Research Chemicals Supplier No. TRCN633500100MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nobiletin
CAS 478-01-3
Purity Grade Notes HPLC
Molecular Formula C21 H22 O8
Quantity 100 mg
Synonym 4H-1-Benzopyran-4-one','2-(3,4-dimethoxyphenyl)-5,6,7,8-tetramethoxy-, Flavone','3',4',5,6,7,8-hexamethoxy- (7CI,8CI), Nobiletin (6CI), 2-(3,4-Dimethoxyphenyl)-5,6,7,8-tetramethoxy-4H-1-benzopyran-4-one, 3',4',5,6,7,8-Hexamethoxyflavone, 5,6,7,8,3',4'-Hexamethoxyflavone, NSC 618903, NSC 76751
InChI Formula InChI=1S/C21H22O8/c1-23-13-8-7-11(9-15(13)24-2)14-10-12(22)16-17(25-3)19(26-4)21(28-6)20(27-5)18(16)29-14/h7-10H,1-6H3
SMILES COc1ccc(cc1OC)C2=CC(=O)c3c(OC)c(OC)c(OC)c(OC)c3O2
IUPAC Name 2-(3,4-dimethoxyphenyl)-5,6,7,8-tetramethoxychromen-4-one
Molecular Weight (g/mol) 402.39
Formula Weight 402.1315
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.