missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-13663
This item is not returnable.
View return policy
Description
NOB1 Polyclonal specifically detects NOB1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| NOB1 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| adenocarcinoma antigen recognized by T lymphocytes 4, ART4, ART-4, MST158, nin one binding protein, NIN1/RPN12 binding protein 1 homolog (S. cerevisiae), NOB1PPSMD8BP1, Phosphorylation regulatory protein HP-10, Protein ART-4, PSMD8 binding protein 1, RNA-binding protein NOB1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| NOB1 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: PSNIKQIQQELEQCDVPEDVRVGCLTTDFAMQNVLLQMGLHVLAVNGMLIREARSYILRCHGCFKTTSDMSRVFCSHCGNKTLKKVSVTVSDDGTLHMHFSR | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 28987 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction