missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NO66 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-93381
This item is not returnable.
View return policy
Description
NO66 Polyclonal specifically detects NO66 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| NO66 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| chromosome 14 open reading frame 169, EC 1.14.11.27, FLJ21802, hsNO66, MAPJD, MYC-associated protein with JmjC domain, NO66lysine-specific demethylase NO66, Nucleolar protein 66 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 79697 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:64000 | |
| NP_078920.2 | |
| RIOX1 | |
| Peptide with sequence C-GAQHSDSKDPR corresponding to internal region according to NP_078920.2. | |
| 0.1 mg | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse and Rat. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction