Learn More
PeproTech NNT1 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: PeproTech 500-P186BT-25UG
Description
AA Sequence of recombinant protein: MLNRTGDPGP GPSIQKTYDL TRYLEHQLRS LAGTYLNYLG PPFNEPDFNP PRLGAETLPR ATVDLEVWRS LNDKLRLTQN YEAYSHLLCY LRGLNRQAAT AELRRSLAHF CTSLQGLLGS IAGVMAALGY PLPQPLPGTE PTWTPGPAHS DFLQKMDDFW LLKELQTWLW RSAKDFNRLK KKMQPPAAAV TLHLGAHGF. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human NNT-1/BCSF-3. Anti-Human NNT-1/BCSF-3-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hNNT-1/BCSF-3 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human NNT-1/BCSF-3 (500-P186) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hNNT-1/BCSF-3. Western Blot: To detect Human NNT-1/BCSF-3 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human NNT-1/BCSF-3 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P186BT-1MG will be provided as 2 x 500 μg
NNT-1/BCSF-3 is a neurotrophic factor with B-cell-stimulating capabilities. Expressed in the lymph nodes and spleen, NNT-1/BCSF-3 activates glycoprotein 130 (gp130) and leukemia inhibitory factor receptor member b (LIFR-b) by binding and inducing the tyrosine phosphorylation of these receptors. In vitro, it supports the survival of chick embryo motor and sympathetic neurons. In mice, NNT-1/BCSF-3 induces serum amyloid A, causes body weight loss, and B cell hyperplasia associated with increases in serum IgG and IgM.
Specifications
| NNT1 | |
| Polyclonal | |
| Biotin | |
| CLCF1 | |
| b cell-stimulating factor 3; B-cell stimulating factor 3; B-cell stimulatory factor 3; B-cell-stimulating factor 3; Bsf3; BSF-3; Cardiotrophin-like cytokine factor 1; CISS2; Clc; CLCF1; CRLF1 associated cytokine-like factor 1; neurotrophin-1/B-cell stimulating factor-3; NNT1; NNT-1; NNT-1/BSF-3; Novel neurotrophin-1; NR6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23529 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| Q9UBD9 | |
| CLCF1 | |
| E.coli-derived, 22.4 kDa Recombinant Human NNT-1/BCSF-3. | |
| 25 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.