missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ nNOS Polyclonal Antibody
GREENER_CHOICE

Product Code. 11571442
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11571442 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11571442 Supplier Invitrogen™ Supplier No. PA1033

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-033 detects nNOS from human, rat and mouse samples. PA1-033 has been successfully used in Western blot, ICC/IF and immunohistochemisty (paraffin) procedures. By Western blot, this antibody detects an ~160 kDa protein. Detects a few non-specific bands in Western blot analysis of rat and mouse brain lysates. This antibody is not recommended for MCF-7, PC-12, SK-N-C or U87-MG cells in Western blot applications. The PA1-033 immunogen is a synthetic peptide to residues C N(1411) R L R S E S I A F I E E S K(1425) of human nNOS. The peptide sequence is 100% conserved in rat, mouse, frog, and rabbit nNos proteins. This peptide (Cat. # PEP-190) is available for use in neutralization and control experiments.

NOS activity requires homodimerization as well as three cosubstrates (L-arginine, NADPH and O2) and five cofactors or prosthetic groups (FAD, FMN, calmodulin, tetrahydrobiopterin and heme). Several distinct NOS isoforms have been described and been shown to represent the products of three distinct genes. These include two constitutive Ca2+/CaM-dependent forms of NOS, including NOS1 (also designated ncNOS) whose activity was first identified in neurons, and NOS3 (also designated ecNOS), first identified in endothelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen nNOS
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene NOS1
Gene Accession No. P29475, P29476, Q9Z0J4
Gene Alias 2310005C01Rik; bNOS; bNOS1; Brain NOS; Constitutive NOS; IHPS1; NC-NOS; neuronal nitric oxide synthase; neuronal nitric oxide synthase NOS1; neuronal NOS; nitric oxidase synthase; nitric oxide synthase; nitric oxide synthase 1; nitric oxide synthase 1 (neuronal); nitric oxide synthase 1, neuronal; nitric oxide synthase, brain; nNOS; N-NOS; nNOS; NOS type I; NO; NOS; NOS Type 1; NOS type I; Nos1; Nos-1; NOS-I; Peptidyl-cysteine S-nitrosylase NOS1; Phospho-bNOS; Phospho-Brain NOS
Gene Symbols NOS1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues C N(1411) R L R S E S I A F I E E S K(1425) of human nNOS.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18125, 24598, 4842
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.