Learn More
NNMT Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15123-1-AP-150UL
Description
The NNMT antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NNMT. This antibody recognizes human, mouse, rat antigen. The NNMT antibody has been validated for the following applications: IF, IP, WB, ELISA analysis.
Specifications
| NNMT | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 4837 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence | |
| 207 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |