missing translation for 'onlineSavingsMsg'
Learn More

NNC 55-0396, MedChemExpress

Product Code. 16440558
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16440558

Brand: Medchem Express HY50722

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NNC 55-0396, Mibefradil derivative, is a highly selective T-type calcium channel blocker; displays IC50 values of 6.8 and > 100 μM for inhibition of Cav3.1 T-type channels and HVA currents respectively in INS-1 cells.

NNC 55-0396, Mibefradil derivative, is a highly selective T-type calcium channel blocker

Specifications

Chemical Name or Material NNC 55-0396
CAS 357400-13-6
Color White
Purity Grade Notes Research
Molecular Formula C30H40Cl2FN3O2
Quantity 5 mg
Synonym NNC 55-0396 dihydrochloride
Solubility Information DMSO : 100 mg/mL (177.13 mM; Need ultrasonic)
SMILES O=C(C1CC1)O[C@@]2(CCN(CCCC3=NC4=CC=CC=C4N3)C)[C@@H](C(C)C)C5=C(C=C(F)C=C5)CC2.[H]Cl.[H]Cl
Molecular Weight (g/mol) 564.56
Formula Weight 564.56
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.