missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NNC 0640

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
NNC 0640 is a negative allosteric modulator of glucagon receptors (pKi = 7.4). Binds to external surface of the transmembrane domain. Also negative allosteric modulator of glucagon-like peptide 1 (GLP-1) receptor. Inhibits GLP-1-mediated cAMP accumulation in vitro.
Specifications
Specifications
| Chemical Name or Material | 4-[[(4-Cyclohexylphenyl)[[[3-(methylsulfonyl)phenyl]amino]carbonyl]amino]methyl]-N-2 H-tetrazol-5-ylbenzamide |
| CAS | 307986-98-7 |
| Quantity | 50 mg |
| Target | Glucagon-Like Peptide 1 Receptor Modulators |
| Molecular Formula | C29H31N7O4S |
| Formula Weight | 8737 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction