missing translation for 'onlineSavingsMsg'
Learn More

NNC-0640, MedChemExpress

Product Code. 16437907
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16437907 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16437907 Supplier Medchem Express Supplier No. HY124622
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress NNC-0640 is a potent human G-protein-coupled glucagon receptor (GCGR) negative allosteric modulator (NAM) with an IC50 of 69.2 nM.

NNC-0640 is a potent human G-protein-coupled glucagon receptor (GCGR) negative allosteric modulator (NAM) with an IC50 of 69.2 nM.

Specifications

Chemical Name or Material NNC-0640
CAS 307986-98-7
Color White
Purity Grade Notes Research
Molecular Formula C29H31N7O4S
Quantity 5 mg
Solubility Information DMSO : 250 mg/mL (435.79 mM; Need ultrasonic)
SMILES O=C(NC1=NN=NN1)C2=CC=C(CN(C3=CC=C(C4CCCCC4)C=C3)C(NC5=CC=CC(S(=O)(C)=O)=C5)=O)C=C2
Molecular Weight (g/mol) 573.67
Formula Weight 573.67
Percent Purity 98.1%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.