missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NNC 0640
GREENER_CHOICE

Product Code. 18715476
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18715476

Brand: Tocris Bioscience™ 6807/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Negative allosteric modulator of glucagon and GLP-1 receptors

NNC 0640 is a negative allosteric modulator of glucagon receptors (pKi = 7.4). Binds to external surface of the transmembrane domain. Also negative allosteric modulator of glucagon-like peptide 1 (GLP-1) receptor. Inhibits GLP-1-mediated cAMP accumulation in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[[(4-Cyclohexylphenyl)[[[3-(methylsulfonyl)phenyl]amino]carbonyl]amino]methyl]-N-2 H-tetrazol-5-ylbenzamide
CAS 307986-98-7
Quantity 10 mg
Target Glucagon-Like Peptide 1 Receptor Modulators
Molecular Formula C29H31N7O4S
Formula Weight 2172
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.