missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NNC 05-2090 hydrochloride, Tocris Bioscience™
GABA uptake inhibitor; moderately BGT-1 selective
Chemical Identifiers
CAS | 184845-18-9 |
---|---|
Molecular Formula | C27H31ClN2O2 |
Molecular Weight (g/mol) | 451.007 |
InChI Key | WDFXPKKFSNMGOO-UHFFFAOYSA-N |
Synonym | nnc 05-2090 hydrochloride, 1-3-9h-carbazol-9-yl propyl-4-2-methoxyphenyl-4-piperidinol hydrochloride, 1-3-9h-carbazol-9-yl-1-propyl-4-2-methoxyphenyl piperidin-4-ol hydrochloride, 1-3-9h-carbazol-9-yl propyl-4-2-methoxyphenyl piperidin-4-ol-hydrogen chloride 1/1 |
PubChem CID | 18782760 |
IUPAC Name | 1-(3-carbazol-9-ylpropyl)-4-(2-methoxyphenyl)piperidin-4-ol;hydrochloride |
SMILES | COC1=CC=CC=C1C2(CCN(CC2)CCCN3C4=CC=CC=C4C5=CC=CC=C53)O.Cl |
Description
GABA uptake inhibitor that displays moderate selectivity for BGT-1 (mGAT-2) transporters (Ki values are 1.4, 15, 19 and 41 μM for hBGT-1, hGAT-3, hGAT-1 and hGAT-2 respectively). Anticonvulsive; inhibits sound-induced tonic and clonic convulsions in DBA/2 mice. Also displays affinity at α1- and D2-receptors (IC50 values are 266 and 1632 nM respectively).Chemical Identifiers
184845-18-9 | |
451.007 | |
nnc 05-2090 hydrochloride, 1-3-9h-carbazol-9-yl propyl-4-2-methoxyphenyl-4-piperidinol hydrochloride, 1-3-9h-carbazol-9-yl-1-propyl-4-2-methoxyphenyl piperidin-4-ol hydrochloride, 1-3-9h-carbazol-9-yl propyl-4-2-methoxyphenyl piperidin-4-ol-hydrogen chloride 1/1 | |
1-(3-carbazol-9-ylpropyl)-4-(2-methoxyphenyl)piperidin-4-ol;hydrochloride |
C27H31ClN2O2 | |
WDFXPKKFSNMGOO-UHFFFAOYSA-N | |
18782760 | |
COC1=CC=CC=C1C2(CCN(CC2)CCCN3C4=CC=CC=C4C5=CC=CC=C53)O.Cl |
Specifications
184845-18-9 | |
nnc 05-2090 hydrochloride, 1-3-9h-carbazol-9-yl propyl-4-2-methoxyphenyl-4-piperidinol hydrochloride, 1-3-9h-carbazol-9-yl-1-propyl-4-2-methoxyphenyl piperidin-4-ol hydrochloride, 1-3-9h-carbazol-9-yl propyl-4-2-methoxyphenyl piperidin-4-ol-hydrogen chloride 1/1 | |
COC1=CC=CC=C1C2(CCN(CC2)CCCN3C4=CC=CC=C4C5=CC=CC=C53)O.Cl | |
451.007 | |
451.01 | |
NNC 05-2090 hydrochloride |
C27H31ClN2O2 | |
WDFXPKKFSNMGOO-UHFFFAOYSA-N | |
1-(3-carbazol-9-ylpropyl)-4-(2-methoxyphenyl)piperidin-4-ol;hydrochloride | |
18782760 | |
>98% |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature