missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMUR1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14619-1-AP-20UL
This item is not returnable.
View return policy
Description
Receptor for the neuromedin-U and neuromedin-S neuropeptides.Specifications
| NMUR1 | |
| Polyclonal | |
| Unconjugated | |
| NMUR1 | |
| FM 3, FM3, G protein coupled receptor 66, GPC R, GPR66, neuromedin U receptor 1, NMUR1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10316, 14767 | |
| -20°C | |
| Liquid |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O55040, Q9HB89 | |
| NMUR1 | |
| NMUR1 Fusion Protein Ag6235 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction