Learn More
NMUR1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14619-1-AP-150UL
 View more versions of this product
Product Code. 16873344
  • £349.00 / 150µL
Description
Receptor for the neuromedin-U and neuromedin-S neuropeptides.
Specifications
Specifications
NMUR1
Polyclonal
Unconjugated
NMUR1
FM 3, FM3, G protein coupled receptor 66, GPC R, GPR66, neuromedin U receptor 1, NMUR1
Rabbit
Antigen Affinity Chromatography
RUO
10316, 14767
-20°C
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O55040, Q9HB89
NMUR1
NMUR1 Fusion Protein Ag6235
150 μL
Primary
Human, Mouse