missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMUR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530376
This item is not returnable.
View return policy
Description
Recommended positive controls: PC-3, rat testis. Predicted reactivity: Human (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Receptor for the neuromedin-U and neuromedin-S neuropeptides.
Specifications
| NMUR1 | |
| Polyclonal | |
| Unconjugated | |
| NMUR1 | |
| (FM-3); FM3; FM-3; G protein-coupled receptor 66; G protein-coupled receptor FM-3; GPC-R; GPR66; G-protein coupled receptor 66; G-protein coupled receptor FM-3; neuromedin U receptor 1; neuromedin-U receptor 1; NMU1R; NmU-R; NMUR1; NMU-R1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10316, 65276 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9HB89, Q9JJI5 | |
| NMUR1 | |
| Recombinant protein fragment corresponding to a region within amino acids 88 and 376 of Human NMUR1 | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction