missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NMUR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599668
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NMUR1.
Receptor for the neuromedin-U and neuromedin-S neuropeptides.
Specifications
| NMUR1 | |
| Polyclonal | |
| Unconjugated | |
| NMUR1 | |
| (FM-3); FM3; FM-3; G protein-coupled receptor 66; G protein-coupled receptor FM-3; GPC-R; GPR66; G-protein coupled receptor 66; G-protein coupled receptor FM-3; neuromedin U receptor 1; neuromedin-U receptor 1; NMU1R; NmU-R; NMUR1; NMU-R1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10316, 14767, 65276 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O55040, Q9HB89, Q9JJI5 | |
| NMUR1 | |
| A synthesized peptide derived from human NMUR1(Accession Q9HB89), corresponding to amino acid residues S9-L59. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction