missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMU, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB25221.100uL
This item is not returnable.
View return policy
Specifications
| NMU | |
| Polyclonal | |
| Unconjugated | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| - | |
| Rabbit | |
| 100 μL | |
| Primary | |
| NMU polyclonal antibody detects endogenous levels of NMU protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| neuromedin U | |
| NMU | |
| A synthetic peptide corresponding to NMU. | |
| RUO | |
| 10874 | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction