missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMT2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32244
This item is not returnable.
View return policy
Description
NMT2 Polyclonal specifically detects NMT2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| NMT2 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| EC 2.3.1.97, glycylpeptide N-tetradecanoyltransferase 2, Myristoyl-CoA:protein N-myristoyltransferase 2, NMT 2, N-myristoyltransferase 2, Peptide N-myristoyltransferase 2, Type II N-myristoyltransferase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:10000, Immunoprecipitation | |
| NP_004799.1 | |
| NMT2 | |
| The immunogen this antibody was made to, maps to a region between residue 100 to 150 of human N-myristoyltransferase 2 using the numbering given in entry NP_004799.1 (GeneID 9397). | |
| 100 μL | |
| Cancer | |
| 9397 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction