missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMT2 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97228B
This item is not returnable.
View return policy
Description
NMT2 Polyclonal antibody specifically detects NMT2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| NMT2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Biotin | |
| EC 2.3.1.97, glycylpeptide N-tetradecanoyltransferase 2, Myristoyl-CoA:protein N-myristoyltransferase 2, NMT 2, N-myristoyltransferase 2, Peptide N-myristoyltransferase 2, Type II N-myristoyltransferase | |
| Produced in rabbits immunized with E. coli-derived Human NMT2 fragment. | |
| 0.1 mL | |
| Cancer | |
| 9397 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction