missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMT1 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32168PE
This item is not returnable.
View return policy
Description
NMT1 Polyclonal antibody specifically detects NMT1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, KnockDown
Specifications
| NMT1 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Knockdown Validated | |
| alternative, short form NMT-S, glycylpeptide N-tetradecanoyltransferase 1, long form, NMT-L, Myristoyl-CoA:protein N-myristoyltransferase 1, NMT 1, NMTEC 2.3.1.97, N-myristoyltransferase 1, Peptide N-myristoyltransferase 1, Type I N-myristoyltransferase | |
| The immunogen this antibody was made to, maps to a region between residue 68 to 118 of human N-Myristoyltransferase 1 using the numbering given in entry NP_066565.1 | |
| 0.1 mL | |
| Cancer | |
| 4836 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunocytochemistry, Immunoprecipitation, KnockDown | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction