missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMRAL1 Polyclonal Antibody

NMRAL1 Polyclonal Antibody
Brand: Invitrogen PA5100642
This item is not returnable.
View return policy
Description
NMRAL1 Polyclonal Antibody for Western Blot
Specifications
| NMRAL1 | |
| Polyclonal | |
| Unconjugated | |
| Nmral1 | |
| 1110025F24Rik; AI256624; HSCARG; LOW QUALITY PROTEIN: nmrA-like family domain-containing protein 1; NmrA like redox sensor 1; NMRAL1; nmral1 {ECO:0000250; NmrA-like family domain containing 1; nmrA-like family domain-containing protein 1; nmrA-like family domain-containing protein 1 {ECO:0000250; RGD1311451; SDR48A1; short chain dehydrogenase/reductase family 48A, member 1; UniProtKB:Q9HBL8} | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 287063, 57407, 67824 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P86172, Q8K2T1, Q9HBL8 | |
| Nmral1 | |
| A synthesized peptide derived from human NMRAL1(Accession Q9HBL8), corresponding to amino acid residues L159-G209. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction