missing translation for 'onlineSavingsMsg'
Learn More

NMNAT-2 Antibody (2E4), Novus Biologicals™

Product Code. 18375968 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375968 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18375968 Supplier Novus Biologicals Supplier No. H00023057M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NMNAT-2 Monoclonal antibody specifically detects NMNAT-2 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMNAT-2
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2E4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_055854
Gene Alias C1orf15, chromosome 1 open reading frame 15, EC 2.7.7.1, EC 2.7.7.18, KIAA0479MGC2756, NaMN adenylyltransferase 2, nicotinamide mononucleotide adenylyltransferase 2, nicotinamide nucleotide adenylyltransferase 2, Nicotinate-nucleotide adenylyltransferase 2, NMN adenylyltransferase 2, PNAT2, pyridine nucleotide adenylyltransferase 2
Host Species Mouse
Immunogen NMNAT2 (NP_055854, 208 a.a. ~ 307 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. CIPGLWNEADMEVIVGDFGIVVVPRDAADTDRIMNHSSILRKYKNNIMVVKDDINHPMSVVSSTKSRLALQHGDGHVVDYLSQPVIDYILKSQLYINASG
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23057
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.