missing translation for 'onlineSavingsMsg'
Learn More

NMNAT-1 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18406146 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406146 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406146 Supplier Novus Biologicals Supplier No. NBP232107FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NMNAT-1 Polyclonal antibody specifically detects NMNAT-1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMNAT-1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.7.1, EC 2.7.7.18, NaMN adenylyltransferase 1, nicotinamide mononucleotide adenylyltransferase 1, nicotinamide nucleotide adenylyltransferase, nicotinamide nucleotide adenylyltransferase 1, Nicotinate-nucleotide adenylyltransferase 1, NMN adenylyltransferase 1, NMNATPNAT1, pyridine nucleotide adenylyltransferase 1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 229 to 279 of human Nicotinamide Nucleotide Adenylyltransferase 1 using the numbering given in entry NP_073624.2 (GeneID 64802).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 64802
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.