missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NMI Recombinant Rabbit Monoclonal Antibody (24GB190)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557125
This item is not returnable.
View return policy
Description
NMI Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
NMYC interactor (NMI) encodes a protein that interacts with NMYC and CMYC (two members of the oncogene Myc family), and other transcription factors containing a Zip, HLH, or HLH-Zip motif. The NMI protein also interacts with all STATs except STAT2 and augments STAT-mediated transcription in response to cytokines IL2 and IFN-gamma. The NMI mRNA has low expression levels in all human fetal and adult tissues tested except brain and has high expression in cancer cell line-myeloid leukemias.
Specifications
| NMI | |
| Recombinant Monoclonal | |
| Unconjugated | |
| NMI | |
| N-mcy (and STAT) interactor; Nmi; N-myc (and STAT) interactor; N-myc and STAT interactor; N-myc interactor; N-myc-interactor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9111 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB190 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13287 | |
| NMI | |
| A synthesized peptide derived from human NMI (aa 1-51). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction