missing translation for 'onlineSavingsMsg'
Learn More

NMI 8739, MedChemExpress

Product Code. 16463408
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463408 10 mg 10mg
16436137 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16463408 Supplier Medchem Express Supplier No. HY101540

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NMI 8739 is a dopamine D2 autoreceptor agonist, which is an amine conjugate of the DHA carrier and the neurotransmitter dopamine.

NMI 8739 is a dopamine D2 autoreceptor agonist, which is an amine conjugate of the DHA carrier and the neurotransmitter dopamine.

Specifications

Chemical Name or Material NMI 8739
CAS 129024-87-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C30H41NO3
Quantity 10 mg
Solubility Information Ethanol : 100 mg/mL (215.68 mM; Need ultrasonic) ∣DMSO : ≥ 100 mg/mL (215.68 mM)
SMILES CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CCC(NCCC1=CC=C(O)C(O)=C1)=O
Molecular Weight (g/mol) 463.65
Formula Weight 463.65
Percent Purity 97.53%
Grade Research
Physical Form Liquid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.