missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NME7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570735
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.
Specifications
| NME7 | |
| Polyclonal | |
| Unconjugated | |
| NME7 | |
| CFAP67; cilia and flagella associated protein 67; D530024H21Rik; MN23H7; NDK 7; NDK7; NDP kinase 7; nm23-H7; Nm23-M7; Nm23-r7; NME/NM23 family member 7; Nme7; non-metastatic cells 7, protein expressed in (nucleoside-diphosphate kinase); nucleoside diphosphate kinase 7; nucleoside diphosphate kinase 7-like protein; nucleoside-diphosphate kinase; nucleoside-diphosphate kinase 7; nucleoside-diphosphate kinase 7 isoform a | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 171567 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9QXL8 | |
| NME7 | |
| synthetic peptide directed towards the N-terminal region of mouse NME7. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction