missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NME7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-42888
This item is not returnable.
View return policy
Description
Specifically detects NME7 in Human samples, and it is validated for Western Blotting
NME7 Polyclonal specifically detects NME7 in Human samples. It is validated for Western Blot.
Specifications
| NME7 | |
| Polyclonal | |
| Western Blot 1:500-1:3000 | |
| EC 2.7.4.6, FLJ37194, MN23H7, NDK 7, NDP kinase 7, nm23-H7, non-metastatic cells 7, protein expressed in (nucleoside-diphosphate kinase), nucleoside diphosphate kinase 7, nucleoside-diphosphate kinase 7 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 29922 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| NME7 | |
| The immunogen used to generate this antibody corresponds to mouse nm23-H7. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction