missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NME6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10250-1-AP-20UL
This item is not returnable.
View return policy
Description
Nucleoside diphosphate (NDP) kinases (EC 2.7.4.6), such as NME6, are ubiquitous enzymes that catalyze transfer of gamma-phosphates, via a phosphohistidine intermediate, between nucleoside and dioxynucleoside tri- and diphosphates.Specifications
| NME6 | |
| Polyclonal | |
| Unconjugated | |
| NME6 | |
| IPIA ALPHA, NDK 6, NDP kinase 6, NME6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10201, 54369, 58964 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O75414, O88425, O88426 | |
| Nme6 | |
| NME6 Fusion Protein Ag0315 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction