missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NME5 Monoclonal Antibody (4F5)
NME5 Monoclonal Antibody (4F5)
Brand: Invitrogen MA520379
This item is not returnable.
View return policy
Description
Specifications
| NME5 | |
| Monoclonal | |
| 0.2 to 1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P56597 | |
| NME5 | |
| NME5 (NP_003542, 121 a.a. ~ 212 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunoprecipitation, Western Blot | |
| 4F5 | |
| Unconjugated | |
| NME5 | |
| 1700019D05Rik; expressed in non-metastatic cells 5; inhibitor of p53-induced apoptosis-beta; IPIA-beta; NDK-H 5; NDP kinase homolog 5; NM23H5; NM23-H5; nm23-M5; NME/NM23 family member 5; NME5; non-metastatic cells 5 protein expressed in; non-metastatic cells 5, protein expressed in (nucleoside-diphosphate kinase); nucleoside diphosphate kinase homolog 5; radial spoke 23 homolog; RSPH23; testis-specific nm23 homolog | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8382 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction