missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NME5 Antibody (4F5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00008382-M04
This item is not returnable.
View return policy
Description
NME5 Monoclonal antibody specifically detects NME5 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| NME5 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| Inhibitor of p53-induced apoptosis-beta, IPIA-beta, NDK-H 5, NDP kinase homolog 5, nm23-H5, NM23H5, non-metastatic cells 5, protein expressed in (nucleoside-diphosphate kinase), nucleoside diphosphate kinase homolog 5, radial spoke 23 homolog, RSPH23, Testis-specific nm23 homolog | |
| NME5 (NP_003542, 121 a.a. ~ 212 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DLRNALHGSNDFAAAEREIRFMFPEVIVEPIPIGQAAKDYLNLHIMPTLLEGLTELCKQKPADPLIWLADWLLKNNPNKPKLCHHPIVEEPY | |
| 0.1 mg | |
| Apoptosis | |
| 8382 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Immunoprecipitation | |
| 4F5 | |
| Western Blot, ELISA, Immunoprecipitation | |
| NP_003542 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction