missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NME3 Monoclonal antibody specifically detects NME3 in Human samples. It is validated for Western Blot, ELISA, ELISA
Specifications
Specifications
| Antigen | NME3 |
| Applications | Western Blot, ELISA, Sandwich ELISA |
| Classification | Monoclonal |
| Clone | 3G10 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_002504 |
| Gene Alias | c371H6.2, DR-nm23NDPK-C, EC 2.7.4.6, KIAA0516, NDK 3, NDP kinase 3, NDP kinase C, NDPKC, NM23H3, NM23-H3, non-metastatic cells 3, protein expressed in, nucleoside diphosphate kinase 3, Nucleoside diphosphate kinase C |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?